    Sort by:
  • Search Within Results
Sepabeads® SP70
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Sepabeads* SP70, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 70 angstroms, Form: 250um Beads, S.A. 870m2/g, P.V. 1.5ml/g, Size: 1KG