    Sort by:
Sephadex® G-10
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (IC19525110)
Supplier:  MP Biomedicals
Description:   Sephadex is a beaded gel filtration medium prepared by cross-linking dextran with epichlorohydrin under alkaline conditions.