    Sort by:
  • Search Within Results
Diaion® WT01S (H)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* WT01S(H), ion exchange resin, weakly acidic porous type, 3.0 meq/ml on polymethacrylate, high reaction rate, Color: White, Form: 106-300um Beads, Size: 1KG