    Sort by:
  • Search Within Results
Diaion® WK10 (H)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* WK10(H), ion exchange resin, weakly acidic porous type, 2.5 meq/ml on polymethacrylate, Form: 300-1180um Beads, Size: 1KG