You Searched For: Diaion® WA20
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46918-A1)
Supplier: Thermo Scientific Chemicals
Description: Diaion* WA20, ion exchange resin, weakly basic porous type, 2.5 meq/ml on poly(styrene-divinylbenzene), Color: Pale yellow, Form: 300-1180um Beads, Size: 1KG