    Sort by:
  • Search Within Results
Diaion® UBK10 (Na)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* UBK10(Na), ion exchange resin, 10% cross-linked, strongly acidic gel type, 2.2 meq/ml on PS-DVB, Form: 560-600um Beads, Size: 1KG