    Sort by:
  • Search Within Results
Diaion® SK112 (Na)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* SK112(Na), ion exchange resin, 12% cross-linked, strongly acidic gel type, 2.1 meq/ml on PS-DVB, Form: 300-1180um Beads, Size: 1KG