    Sort by:
  • Search Within Results
Diaion® SK104 (Na)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* SK104(Na), ion exchange resin, 4% cross-linked, strongly acidic gel type, 1.2 meq/ml on PS-DVB, Color: Brown, Form: 300-1180um Beads, Size: 1KG