    Sort by:
  • Search Within Results
Diaion® SA11A (Cl)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* SA11A(Cl), ion exchange resin, lightly cross-linked, strongly basic gel type I, 0.85 meq/ml on PS-DVB, Color: White to yellow, Form: 300-1180um Beads, Size: 1KG