    Sort by:
  • Search Within Results
You Searched for: "diaion-rcp160m-h"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* RCP160M(H), ion exchange resin, highly cross-linked, strongly acidic highly porous type, 1.5 meq/ml on PS-DVB, Color: Dark brown, Form: 250-710um Beads, Size: 1KG