    Sort by:
You Searched For:
Diaion® PK220 (Na)
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* PK220(Na), ion exchange resin, 10% cross-linked, strongly acidic porous type, 1.9 meq/ml on PS-DVB, Form: 300-1180um Beads, Size: 1KG