You Searched For: Diaion® PA418 (Cl)
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46837-A1)
Supplier: Thermo Scientific Chemicals
Description: Diaion* PA418(Cl), ion exchange resin, 9% cross-linked, strongly basic porous type II, 1.3 meq/ml on PS-DVB, Cas Number: 69011-15-0, Color: Pale yellow, Form: 300-1180um Beads, Size: 1KG