    Sort by:
  • Search Within Results
You Searched for: "diaion-pa312-cl"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* PA312(Cl), ion exchange resin, 6% cross-linked, strongly basic porous type I, 1.2 meq/ml on PS-DVB, Form: 300-1180um Beads, Size: 1KG