You Searched For: Diaion® PA308 (Cl)
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46990-A1)
Supplier: Thermo Scientific Chemicals
Description: Diaion* PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Color: White to yellow, Form: 300-1180um Beads, Size: 1KG