    Sort by:
  • Search Within Results
Diaion® NSA100 (Cl)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* NSA100(Cl), ion exchange resin, highly cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB, Color: White to yellow, Form: 300-1180um Beads, Size: 1KG