You Searched For: Diaion® HPA25 (Cl)
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46691-A1)
Supplier: Thermo Scientific Chemicals
Description: Diaion* HPA25(Cl), ion exchange resin, highly cross-linked, strongly basic highly porous type I, 0.5 meq/ml on PS-DVB, Form: 250um Beads, Size: 1KG