    Sort by:
  • Search Within Results
Diaion® CRB03 (OH)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* CRB03(OH), chelating resin, highly porous type, 0.7 meq/ml on poly(styrene-divinylbenzene), Form: 300-1180um Beads, Size: 1KG