    Sort by:
You Searched For:
Diaion® CR20
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* CR20, chelating resin, highly porous type, 0.4 mmole/ml on poly(styrene-divinylbenzene), Color: Pale yellow, Form: 300-1180um Beads, Size: 1KG