    Sort by:
  • Search Within Results
Diaion® CR11 (Na)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Diaion* CR11(Na), chelating resin, highly porous type, 0.5 mmole/ml on poly(styrene-divinylbenzene), Color: Pale yellow, Form: 355-1180um Beads, Size: 1KG