    Sort by:
  • Search Within Results
D.E.R.™ 732 Resin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (100504-848)
Supplier:  Electron Microscopy Sciences
Description:   Used as a flexibilizer in Maraglas 655-epoxy formulation. Relatively low Viscosity (55 to 100 at 25 °C).
Minority or Woman-Owned Business Enterprise