You Searched For: 1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 1,1-Bis((2R,5R)-2,5-di-i-propylphospholano)ferrocene, Purity: 97% 99%ee, CAS Number: 849950-54-5, Appearance: Yellow Powder, Storage: Inert atmosphere, 2-8 C, Size: 5g