    Sort by:
  • Search Within Results
3,5-Dichlorophenylmagnesium bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   3,5-Dichlorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal®, Cas number: 82297-90-3, Molecular Formula: C6 H3 Br Cl2 Mg, Molecular Weight: 250.2, Form: Brown to black, Liquid, Size: 50ML