    Sort by:
  • Search Within Results
4-Methoxy-2-methylphenylmagnesium bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   4-Methoxy-2-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal®, Cas number: 148706-30-3, Molecular Formula: C8 H9 Br Mg O, Molecular Weight: 225.37, Form: Clear colorless to brown, Solution, Size: 50ML