    Sort by:
  • Search Within Results
4-Isopropylphenylmagnesium bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   4-Isopropylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal®, Cas number: 18620-03-6, Molecular Formula: C9 H11 Br Mg, Molecular Weight: 223.4, Form: Yellow to brown, Solution, Size: 50ML