    Sort by:
  • Search Within Results
5-Bromo-1-methyl-1H-indole-3-carboxylic acid
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   5-Bromo-1-methyl-1H-indole-3-carboxylic acid, Purity: 97%, Cas number: 400071-95-6, Molecular Formula: C10 H8 Br N O2, Molecular Weight: 254.08, Appearance: White to off-white Crystalline powder, Size: 1G