    Sort by:
  • Search Within Results
Diethyl-3,4-pyridinedicarboxylate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (103514-292)
Supplier:  Acros Organics
Description:   Diethyl 3,4-pyridinedicarboxylate, purity: 97%, CAS Number: 1678-52-0, Molecular Formula: C11H13NO4, Molecular Wight: 223.23, Synonyms: Cinchomeronic ester, Form: Liquid, color: Clear colorless to yellow, size: 5G