    Sort by:
  • Search Within Results
1,2-Dibromocyclopentene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   1,2-Dibromocyclopentene, purity: 97%, CAS Number: 75415-78-0, Molecular Formula: C5H6Br2, Molecular Wight: 225.90, Form: Liquid, color: Clear colorless to yellow, size: 1G