    Sort by:
  • Search Within Results
Di-n-butylmagnesium
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   Di-n-butylmagnesium 0.5M solution in heptane, AcroSeal*, Cas number: 1191-47-5, Molecular Weight: 138.54, Molecular formula: C8 H18 Mg, form: Solution, Color: Clear to slightly hazy colorless, Size: 100ML