    Sort by:
  • Search Within Results
(Trimethylsilyl)methyllithium
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   (Trimethylsilyl)methyllithium 0.7M (10 wt%) solution in hexanes, AcroSeal*, Cas number: 1822-00-0, Molecular Weight: 94.16, Molecular formula: C4 H11 Li Si, form: Liquid, Color: Colorless to yellow-brown, Size: 800ML