    Sort by:
  • Search Within Results
3,4-Di-O-acetyl-L-arabinal
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (103465-096)
Supplier:  Acros Organics
Description:   3, 4-Di-O-acetyl-L-arabinal, Purity: 97%, CAS number: 3945-18-4, Molecular Formula: C9H12O5Infrared SpectrumAuthenticBeilstein17, IV, 2035Refractive Index1.4580 to 1.4600 (20 deg C, 589nm), Color: Colorless to yellow, Size: 1G