    Sort by:
  • Search Within Results
Dimethyl-trans-1,2-cyclopropanedicarboxylate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   Dimethyl trans-1,2-cyclopropanedicarboxylate, 99+%, CAS number: 826-35-7, Molecular Formula: C7 H10 O4, Molecular weight: 158.16, Color: Clear colorless, Form: Liquid, Size: 5G