    Sort by:
  • Search Within Results
4-(Dimethylamino)phenethyl alcohol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   4-(Dimethylamino)phenethyl alcohol, Purity: 99%, CAS Number: 50438-75-0, Molecular Formula: C10 H15 N O, Molecular weight: 165.23, Synonyms: 2-[4-(Dimethylamino)phenyl]ethanol, Form: Crystalline Powder, Color: Light brown, Size: 1g