    Sort by:
  • Search Within Results
4,4'-Dimethoxystilbene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (103509-258)
Supplier:  Acros Organics
Description:   4,4'-Dimethoxystilbene, Cas number: 4705-34-4, Purity: 99%, Molecular Weight: 240.30, Molecular Formula: C16 H16 O2, Form: Solid, Color: White, Size: 10G