    Sort by:
  • Search Within Results
Gum guaiac
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   Soluble in alcohol, chloroform, ether and menthanol (10 mg/ mL-hazy solution); slightly soluble in benzene and carbon disulfide; insoluble in water.