    Sort by:
  • Search Within Results
Acid blue 29
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   Slightly soluble in methyl cellosolve and water (20 mg/mL); very slightly soluble in ethanol (2 mg/mL).