    Sort by:
  • Search Within Results
γ-L-Glutamyl-p-nitroanilide hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   Soluble in water (5 mg/mL - clear, light yellow solution) or 0.1 N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100 mg/mL with sligh...