You Searched For: Desmosine
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: MP Biomedicals
Description: Desmosine is prepared according to the method of Starcher et al. from hydrolysed neck ligament by solvent extraction and chromatography on Dowex 50.