    Sort by:
  • Search Within Results
4-(1,4-Cyclohexadien-1-yl)-1-butanol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   4-(1, 4-Cyclohexadien-1-yl)-1-butanol, Purity: 97%, stab. with 0.1%, BHTMolecular formula: C10H16O, Synonym: 4-Cyclohexa-1, 4-dienyl-1-butanol, Form: Liquid, Color: Colorless, size: 1g