    Sort by:
  • Search Within Results
3-Chloro-2-fluorobenzylmagnesium bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   3-Chloro-2-fluorobenzylmagnesium bromide, 0.25M in 2-Methyltetrahydrofuran, CAS Number: 1268684-42-9, Molecular Formula: C7H5BrClFMg, Color: Grey, Form: Liquid, Size: 100ml