    Sort by:
  • Search Within Results
(S)-An-phanephos
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (AAH35716-03)
Supplier:  Alfa Aesar
Description:   (S)-An-Phanephos, Molecular formula: C44H42O4P2, Synonym: (S)-4, 12-Bis[di(4-methoxyphenyl)phosphino][2.2]paracyclophane, Form: Solid, Color: White, size: 1g