    Sort by:
  • Search Within Results
(1,3-Dioxolan-2-ylmethyl)zinc bromide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   (1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, Molecular Formula: C4H7BrO2Zn, Color: yellow to brown to black, Form: liquid, packaged under Argon in resealable ChemSeal bottles, Size: 50ML