Spebrutinib

Synonyms: N-(3-((5-Fluoro-2-((4-(2-methoxyethoxy)phenyl)amino)pyrimidin-4-yl)amino)phenyl)acrylamideAVL-292

9413-5 9413-25
76022-046EA 99.08 USD
76022-046 76022-102
Spebrutinib
Spebrutinib

AVL-292 is a potent, selective inhibitor of Bruton’s Tyrosine Kinase (BTK) (IC₅₀ = 0


5 nM) with potential antineoplastic activity.

Formula: C₂₂H₂₂FN₅O₃
MW: 423,44 g/mol
MDL Number: MFCD25976876
CAS Number: 1202757-89-8
https://us.vwr.com/stibo/web/std.lang.all/23/96/23492396.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR