I-CBP112

Supplier: BioVision

AB284903-5MG AB284903-1MG
76022-014EA 137.91 USD
76022-014 76022-016
I-CBP112
I-CBP112

I-CBP112 is a selective CBP/EP300 bromodomain inhibitor (IC₅₀ values of 170 nM, 170 nM, 151 nM and 625 nM for CREBBP(Alphascreen), CREBBP(BLI), CREBBP(ITC) and EP300(ITC),respectively)


Selective for CBP and p300 over ATAD2, BAZ2B, BRD2(2), BRD4(1), PB1(5), PCAF, PHIP(2) and TIF1α bromodomains in a BLI assay.

Formula: C₂₇H₃₆N₂O₅
MW: 468.59 g/mol
CAS Number: 1640282-31-0
https://us.vwr.com/stibo/web/std.lang.all/23/89/23492389.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR