XL-147

9401-25 9401-5
76021-936EA 192.64 USD
76021-936 76021-938
XL-147
XL-147

XL-147 is an ATP-competitive, highly selective, and reversible pan-class I phosphoinositide 3-kinase (PI3K) inhibitor


It inhibits PI3K p110α, -β, -γ, and -δ isoforms, with IC₅₀ values of 48, 617, 10, and 260 nmol/L, respectively, measured in biochemical assays. XL-147 displays antitumor activity.

Formula: C₂₁H₁₆N₆O₂S₂
MW: 448,52 g/mol
MDL Number: MFCD16621245
CAS Number: 956958-53-5
https://us.vwr.com/stibo/web/std.lang.all/23/83/23492383.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR