Polyvinylpyrrolidone

Supplier: BIOASSAY SYSTEMS

Synonyms: PolyvidonePolyvinylpyrrolidonPVP

PVPP-10
76121-356EA 34.3 USD
76121-356
Polyvinylpyrrolidone
Polyvinylpyrrolidone

Polyvinylpyrrolidone (PVPP) used for preparation of wine samples for use with the QuantiQuik™ Histamine Quick Test Strips.


  • Powder

Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR