Balicatib

Synonyms: N-(1-((Cyanomethyl)carbamoyl)cyclohexyl)-4-(4-propylpiperazin-1-yl)benzamide

2596-5 2596-25
76021-170EA 110.91 USD
76021-170 76021-172
Balicatib
Balicatib

Cell-permeable


Balicatib is a potent human cathepsin K inhibitor in vitro (IC₅₀ = 1.4 nM) with a high selectivity against human cathepsins B, L, and S (> 4,800-fold, > 500-fold and > 65,000-fold, respectively).

Formula: C₂₃H₃₃N₅O₂
MW: 411,54 g/mol
MDL Number: MFCD19443790
CAS Number: 354813-19-7
https://us.vwr.com/stibo/web/std.lang.all/21/21/23492121.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR