Evans blue

2586-1G
76021-136EA 81.6 USD
76021-136
Evans blue
Evans blue

Evans Blue is a potent inhibitor of the uptake of L-glutamate into synaptic vesicles


Evans blue also inhibits AMPA and kainate receptor-mediated currents with IC₅₀ values of 220 and 150 nM respectively. Also acts as a P2X-selective purinoceptor antagonist.

Formula: C₃₄H₂₈N₆O₁₄S₄
MDL Number: MFCD00004021
CAS Number: 314-13-6
https://us.vwr.com/stibo/web/std.lang.all/21/10/23492110.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR