WP1066, EZSolution™

Supplier: BioVision

Synonyms: (S,E)-3-(6-Bromopyridin-2-yl)-2-cyano-N-(1-phenylethyl)acrylamide

AB146607-5MG
76020-940EA 255.88 USD
76020-940
WP1066, EZSolution™
WP1066

A potent, cell-permeable JAK2 inhibitor


A novel analog of the JAK2 inhibitor AG490 (Cat. No. 1570-5) that blocks STAT3 and phosphoinositide-3-kinase pathways. It is significantly more potent and active against human malignant glioma cells in vitro and in vivo than AG 490. Similar to AG490, WP1066 inhibits the phosphorylation of JAK2, but unlike AG490, WP1066 also degrades JAK2 protein, thus blocking its downstream signal transducer and activator of transcription (STAT) and phosphoinositide-3-kinase pathways to results in the activation of the caspase pathway.

Formula: C₁₇H₁₄BrN₃O
MW: 356,22 g/mol
MDL Number: MFCD12912450
CAS Number: 857064-38-1
https://us.vwr.com/stibo/web/std.lang.all/20/70/23492070.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR