TGR5-Receptor-Agonist

Synonyms: TGR5TGR5-Receptor-AgonistMDK-00245MDK00245MDK 002453-(2-Chlorophenyl)-N-(4-chlorophenyl)-N,5-dimethylisoxazole-4-carboxamide3-(2-chlorophenyl)-N-(4-chlorophenyl)-N,5-dimethyl-4-isoxazolecarboxamide

2538-5
76020-920EA 297.72 USD
76020-920
TGR5-Receptor-Agonist
TGR5-Receptor-Agonist

Cell-permeable, small molecule agonist of TGR5 G-protein coupled receptor


This agonist has shown to increase glucagon like peptide-1 (GLP-1) secretion from primary intestinal cells.

Formula: C₁₈H₁₄Cl₂N₂O₂
MW: 361.22 g/mol
MDL Number: MFCD17169972
CAS Number: 1197300-24-5
https://us.vwr.com/stibo/web/std.lang.all/20/61/23492061.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR