Artemisinin

Synonyms: QinghaosuArteannuin

2519-250 2519-50
76020-812EA 79.26 USD
76020-812 76020-814
Artemisinin
Artemisinin

Artemisinin is a sesquiterpene natural product originally isolated from plants of the genus Artemisia


It effectively kills malarial parasites of the genus Plasmodium (IC₅₀ = 4.0 nM). Displays antiangiogenic effects in mouse embryonic stem cell-derived embryoid bodies. Has potential applications in certain types of cancer and inflammatory conditions.

Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
MDL Number: MFCD00081057
CAS Number: 63968-64-9
https://us.vwr.com/stibo/web/std.lang.all/20/44/23492044.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR